Please login to your online account to display your discounted pricing

Ammonium fluoride, +98%, extra pure, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  201845000

 View more versions of this product

Catalog No. AC201845000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.0% (Alkalimetry)
Purity Grade Extra Pure
Physical Form Adhering Crystals and/or Chunks
Formula Weight 37.04
Color Colorless or white to off-white
Quantity 500g
Merck Index 15, 519
TSCA TSCA
RTECS Number BQ6300000
EINECS Number 235-185-9
Heavy Metals (as Pb) 5ppm max.
Packaging Plastic bottle
Chemical Name or Material Ammonium fluoride
Assay Percent Range 98+%
Sulfate 50ppm max.
MDL Number MFCD00011423
CAS 12125-01-8
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Hexafluorosilicate 0.1 % max.
Iron (Fe) 5ppm max.
Name Note p.a.
Residue after Ignition 0.01 % max. (SO4)
Ammonium Bifluoride 1 % max.
Molecular Formula H4FN
Linear Formula NH4F
Chlorine (Cl) 10ppm max.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.

Specifications

Chemical Name or Material Ammonium fluoride
Name Note p.a.
CAS 12125-01-8
Purity Grade Extra Pure
Assay ≥98.0% (Alkalimetry)
Assay Percent Range 98+%
Molecular Formula H4FN
Linear Formula NH4F
Formula Weight 37.04
MDL Number MFCD00011423
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.

SDS & Safety Documents