Learn More
Ammonium fluoride, 98+%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 393192500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium fluoride | |
12125-01-8 | |
250 g | |
H4FN | |
MFCD00011423 | |
Solubility in water: soluble. Other solubilities: slightly soluble in alcohol | |
37.04 | |
Analytical | |
0.82 |
Colorless to Yellow or White to Yellow | |
Liquid or Solid | |
98+% | |
NH4F | |
15, 519 | |
37.04 | |
≥98% | |
Plastic bottle |
Chemical Identifiers
12125-01-8 | |
H4FN | |
37.04 | |
MFCD00011423 |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air
GHS Signal Word: Danger
EINECSNumber : 235-185-9
RUO – Research Use Only