Please login to your online account to display your discounted pricing

Ammonium fluoride, 0.5M solution in methanol, ACROS Organics™


Quantity 100mL
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC393011000

 View more versions of this product

Catalog No. AC393011000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 37.04
Quantity 100mL
Color Clear colorless
Density 0.8200g/mL
Concentration 0.45 to 0.55M
Merck Index 15, 519
Linear Formula NH4F
Chemical Name or Material Ammonium fluoride
MDL Number MFCD00011423
Packaging Plastic bottle
Molecular Formula H4FN
CAS 12125-01-8
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.
CAS 67-56-1
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Ammonium fluoride
CAS 12125-01-8
CAS 67-56-1
Molecular Formula H4FN
Linear Formula NH4F
Formula Weight 37.04
MDL Number MFCD00011423
Accessories

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.

SDS & Safety Documents