missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium fluoride, 0.5M solution in methanol, ACROS Organics™

Chemical Identifiers

CAS 12125-01-8,67-56-1
Molecular Formula H4FN
Linear Formula NH4F
Formula Weight 37.04
MDL Number MFCD00011423
Chemical Name or Material Ammonium fluoride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC393011000 SDS Thermo Fisher Scientific
AC393011000
Plastic bottle 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC393018000 SDS Thermo Fisher Scientific
AC393018000
Plastic bottle 800mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

12125-01-8,67-56-1
NH4F
MFCD00011423
H4FN
37.04
Ammonium fluoride

Specifications

12125-01-8,67-56-1
H4FN
37.04
15, 519
0.8200g/mL
Clear colorless
Ammonium fluoride
Liquid
NH4F
MFCD00011423
0.45 to 0.55M
Plastic bottle
100mL

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.