missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium ferrocyanide hydrate, 90-95%, pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC209730500

 View more versions of this product

Catalog No. AC209730500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Ammonium ferrocyanide hydrate
14481-29-9
Fine Crystalline Powder
90-95%
(NH4)4Fe(CN)6·xH2O
MFCD00149957
Ammonium hexacyanoferrate
suitable for determination of calcium
50g
Pure
Pure
≥90%
C6H16FeN10·xH2O
284.1
15, 517
Glass bottle
Green

Chemical Identifiers

14481-29-9
C6H16FeN10·xH2O
284.1
Ammonium hexacyanoferrate
≥90%
90-95%
(NH4)4Fe(CN)6·xH2O
MFCD00149957
Ammonium ferrocyanide hydrate, Pure
Pure

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.