Please login to your online account to display your discounted pricing

Ammonium ferrocyanide hydrate, 90-95%, pure, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ammonium ferrocyanide hydrate
Name Note Pure
CAS 14481-29-9
Purity Grade Pure
Assay ≥90%
Assay Percent Range 90-95%
Molecular Formula C6H16FeN10·xH2O
Linear Formula (NH4)4Fe(CN)6·xH2O
Formula Weight 284.1
MDL Number MFCD00149957
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC209730500

  SDS
thermo fisher scientific
AC209730500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fine Crystalline Powder
Formula Weight 284.1
Assay ≥90%
Quantity 50g
Color Green
Purity Grade Pure
Merck Index 15, 517
Packaging Glass bottle
Linear Formula (NH4)4Fe(CN)6·xH2O
Name Note Pure
Molecular Formula C6H16FeN10·xH2O
Chemical Name or Material Ammonium ferrocyanide hydrate
CAS 14481-29-9
Assay Percent Range 90-95%
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
MDL Number MFCD00149957
Suitability suitable for determination of calcium
Synonym Ammonium hexacyanoferrate
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.