missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Dihydrogen Phosphate, Reagent ACS, +98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC423380025

 View more versions of this product

Catalog No. AC423380025

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.0%
Crystalline Solid
2.5kg
3.8 to 4.4 (5% soln. at 25°C)
15, 541
100ppm max.
50ppm max.
MFCD00003396
H6NO4P
NH4H2PO4
10ppm max.
5ppm max.
50ppm max.
115.03
ACS Reagent Grade
White
1.8
0.005% max.
10ppm max.
5ppm max.
Ammonium dihydrogen phosphate
7722-76-1
98+%
5ppm max.
10ppm max.
Ammonium phosphate, monobasic

Chemical Identifiers

7722-76-1
H6NO4P
115.03
Ammonium phosphate, monobasic
≥98.0%
98+%
NH4H2PO4
MFCD00003396
Ammonium dihydrogen phosphate
ACS Reagent Grade

Safety and Handling

SDS & Safety Documents

EINECSNumber : 231-764-5

TSCA : TSCA