missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Dihydrogen Phosphate, MilliporeSigma™

For analysis EMSURE™ ACS, Reag. Ph Eur

Manufacturer:  MilliporeSigma 1011269050

 View more versions of this product

Catalog No. M1011269050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ammonium dihydrogen phosphate
H6NO4P
Soluble in water: 368g/L at 20°C
[NH4+].OP(=O)(O)[O-]
115.025
CHEBI:62982
1.80g/cm3 at 20°C
Fibre carton
0.066hPa at 125°C
7722-76-1
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
LFVGISIMTYGQHF-UHFFFAOYSA-N
azanium;dihydrogen phosphate
24402
115.02
Odorless
ACS, Reag. Ph Eur
50g

Chemical Identifiers

7722-76-1
115.025
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
CHEBI:62982
[NH4+].OP(=O)(O)[O-]
H6NO4P
LFVGISIMTYGQHF-UHFFFAOYSA-N
24402
azanium;dihydrogen phosphate
Safety and Handling

Safety and Handling

EINECSNumber : 231-764-5

Recommended Storage : Store at 5° to 30°C (41° to 86°F), Tightly closed. Dry

Documents
Promotions

Promotions