Please login to your online account to display your discounted pricing

Ammonium dihydrogen phosphate, +99%, for analysis, ACROS Organics™


Quantity 10kg
Packaging Plastic Drum

Manufacturer: thermo fisher scientific  AC201830100

 View more versions of this product

Catalog No. AC201830100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Crystalline Solid
Formula Weight 115.03
Quantity 10kg
Chemical Name or Material Ammonium Dihydrogen Phosphate
Packaging Plastic Drum
Melting Point 190°C
Color White
Purity Grade For analysis
pH 4 to 4.5 (5% soln. at 20°C)
Density 1.8g/mL
Heavy Metals (as Pb) 5ppm max.
Potassium (K) 50ppm max.
Nitrate 10ppm max.
Arsenic (As) 0.5ppm max.
Sulfate 50ppm max.
Chlorine (Cl) 5ppm max.
Molecular Formula H6NO4P
MDL Number MFCD00003396
Assay Percent Range 99+%
Sodium (Na) 50ppm max.
Linear Formula NH4H2PO4
Iron (Fe) 10ppm max.
Synonym Ammonium phosphate, monobasic
CAS 7722-76-1

Specifications

Chemical Name or Material Ammonium Dihydrogen Phosphate
CAS 7722-76-1
Purity Grade For analysis
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula H6NO4P
Linear Formula NH4H2PO4
Formula Weight 115.03
MDL Number MFCD00003396
Synonym Ammonium phosphate, monobasic