missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium dihydrogen phosphate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 436791000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisSpecifications
Ammonium dihydrogen phosphate | |
White or Colorless | |
Crystals | |
99.999% | |
NH4H2PO4 | |
15,541 | |
Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone | |
N.OP(O)(O)=O | |
115.03 | |
CHEBI:62982 | |
99.999% | |
Plastic bottle |
190.0°C | |
7722-76-1 | |
100 g | |
H6NO4P | |
MFCD00003396 | |
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
phosphoric acid amine | |
24402 | |
115.03 | |
Trace Metal Basis |
Chemical Identifiers
7722-76-1 | |
115.03 | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
24402 | |
phosphoric acid amine |
H6NO4P | |
MFCD00003396 | |
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
CHEBI:62982 | |
N.OP(O)(O)=O |
Safety and Handling
EINECSNumber : 231-764-5
RUO – Research Use Only