missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium dihydrogen arsenate, 98%

Manufacturer:  Alfa Aesar™ 3149318

 View more versions of this product

Catalog No. AA3149318



Specifications

Specifications

Ammonium dihydrogen arsenate
98%
MFCD00064864
Ammonium arsenate, monobasic
[NH4+].O[As](=O)(O)[O-]
158.973
158.98
2.311g/mL
300°C (decomposition)
13462-93-6
AsH6NO4
UN1557
VTWJXWNBCACAAK-UHFFFAOYSA-N
azanium;dihydrogen arsorate
166814
Crystalline
1.577
50g

Chemical Identifiers

13462-93-6
158.973
VTWJXWNBCACAAK-UHFFFAOYSA-N
166814
[NH4+].O[As](=O)(O)[O-]
AsH6NO4
MFCD00064864
Ammonium arsenate, monobasic
azanium;dihydrogen arsorate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H350
Toxic if swallowed.
Toxic if inhaled.
May cause cancer.

GHS P Statement
P261-P281-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.

TSCA : TSCA

SDS Documents