Learn More
Ammonium dihydrogen arsenate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03149330
Description
It is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium dihydrogen arsenate | |
13462-93-6 | |
250 g | |
MFCD00064864 | |
ammonium dihydrogen arsenate, ammonium arsenate ortho, ammonium dihydrogenarsenate, unii-0kyl9q413a, arsenic acid h3aso4 , monoammonium salt, acmc-20alsv, azanium dihydrogen arsorate, arsenic acid h3aso4 ,ammonium salt 1:1, arsenic acid h3aso4 , ammonium salt 1:1 | |
VTWJXWNBCACAAK-UHFFFAOYSA-N | |
azanium;dihydrogen arsorate | |
166814 | |
98% | |
2.311 g/mL |
300°C (decomposition) | |
Crystalline | |
AsH6NO4 | |
UN1557 | |
Soluble in water. (337.4mg/ml at 0°C). | |
[NH4+].O[As](=O)(O)[O-] | |
158.973 | |
158.98 | |
1.577 |
Chemical Identifiers
13462-93-6 | |
158.973 | |
VTWJXWNBCACAAK-UHFFFAOYSA-N | |
166814 | |
[NH4+].O[As](=O)(O)[O-] |
AsH6NO4 | |
MFCD00064864 | |
ammonium dihydrogen arsenate, ammonium arsenate ortho, ammonium dihydrogenarsenate, unii-0kyl9q413a, arsenic acid h3aso4 , monoammonium salt, acmc-20alsv, azanium dihydrogen arsorate, arsenic acid h3aso4 ,ammonium salt 1:1, arsenic acid h3aso4 , ammonium salt 1:1 | |
azanium;dihydrogen arsorate |
Safety and Handling
GHS H Statement
H301-H331-H350
Toxic if swallowed.
Toxic if inhaled.
May cause cancer.
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
EINECSNumber : 236-667-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only