Please login to your online account to display your discounted pricing

Ammonium Dichromate (Crystalline/Certified), Fisher Chemical

$174.54 - $3,793.15

Chemical Identifiers

Chemical Name or Material Ammonium Dichromate
CAS 7789-09-5
Purity Grade Certified Grade
Physical Form Solid
Molecular Formula H8Cr2N2O7
Linear Formula (NH4)2Cr2O7
Formula Weight 252.07
MDL Number MFCD00010879
Synonym Chromic Acid Diammonium Salt
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A644-100

  SDS
fisher scientific company
FLA644100
Glass Bottle 100g Each for $174.54
Case of 6 Each for $1,047.24

A644-500

  SDS
fisher scientific company
FLA644500
Glass Bottle 500g Each for $362.62
Case of 6 Each for $2,175.72

A644-212

  SDS
fisher scientific company
FLA644212
Glass Bottle 2.5kg Case of 4 Each for $3,793.15
Description & Specifications

Specifications

Melting Point 170°C
Purity Grade Certified Grade
Formula Weight 252.07
Quantity 100g
DOT Information DOT Class 5.1, : Oxidizer
Physical Form Solid
Color Orange
Molecular Formula H8Cr2N2O7
Density 2.1500g/cm³
pH 3 to 4
Identification Pass Test
Chloride 0.005% max.
Calcium (Ca) 0.002% max.
Packaging Glass Bottle
MDL Number MFCD00010879
Insoluble Matter 0.005% max.
Synonym Chromic Acid Diammonium Salt
Sulfate 0.006% max.
Alkalis and Other Salts 0.15% max.
CAS 7789-09-5
Linear Formula (NH4)2Cr2O7
Chemical Name or Material Ammonium Dichromate
ChemAlert Storage Symbol Yellow

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling