Please login to your online account to display your discounted pricing

Ammonium Dichromate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$245.04 - $969.15

Chemical Identifiers

Chemical Name or Material Ammonium Dichromate
CAS 7789-09-5
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula (NH4)2Cr2O7
Formula Weight 252.07
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002279

 
avantor performance
068805
Poly Bottle 2.5kg Each for $969.15

02002280

 
avantor performance
068801
Poly Bottle 500g Each for $245.04
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 252.07
Chemical Name or Material Ammonium Dichromate
Molecular Formula (NH4)2Cr2O7
CAS 7789-09-5
Packaging Poly Bottle
Specific Gravity 1L = 2.15kg
Quantity 2.5kg