missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium dichromate, 99.5%, Alfa Aesar

Catalysts, source of pure nitrogen, in lithography

Manufacturer:  Alfa Aesar 1344436

 View more versions of this product

Catalog No. AA1344436



Specifications

Specifications

Ammonium dichromate
99.5%
ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
252.06
252.1
500g
7789-09-5
Cr2H8N2O7
JOSWYUNQBRPBDN-UHFFFAOYSA-P
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
24600
Crystalline

Chemical Identifiers

7789-09-5
252.06
ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Cr2H8N2O7
JOSWYUNQBRPBDN-UHFFFAOYSA-P
24600
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
Documents