Learn More
Ammonium diamminetetrathiocyanatochromate(III) monohydrate, ACS, 93.0% min, Thermo Scientific Chemicals
Used to precipitate primary and secondary amines as their ammonium salts
Supplier: Thermo Scientific Chemicals 08777722
Description
Ammonium diamminetetrathiocyanatochromate(III) monohydrate is used to precipitate primary and secondary amines as their ammonium salts. It is a color reagent and precipitant for mercury. It is also used as a precipitant for primary and secondary amines and certain amino acids. It is a reagent used for quantitative studies, including titrimetric, gravimetric, and spechtometric assays. Additionally, Reinecke salt has also been shown to determine the concentration of Dimenhydrinate using a spectrophotometric procedure. Other studies use Reinecke salt as a chemical actinometer to calculate the photodegradation of Manidipine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium diamminetetrathiocyanatochromate (III) monohydrate | |
13573-17-6 | |
100 g | |
NH4[Cr(NH3)2(SCN)4]·H2O | |
UN3077 | |
Reinecke salt; Ammonium reineckate | |
354.44 (336.42 Anhydrous) | |
ACS |
Red | |
Powder | |
C4H10CrN7S4·H2O | |
MFCD00066638 | |
14,8128 | |
Insoluble in dilute hydrochloric acid; Moderately soluble in water; Soluble in alcohol | |
≥93.0% | |
Odorless |
Chemical Identifiers
13573-17-6 | |
C4H10CrN7S4·H2O | |
MFCD00066638 | |
Reinecke salt; Ammonium reineckate |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 237-003-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only