Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium cobalt(II) phosphate monohydrate, 98%

$59.38 - $59.38

Chemical Identifiers

Chemical Name or Material Ammonium cobalt(II) phosphate monohydrate
CAS 16827-96-6
Molecular Formula H4CoNO4P·H2O
Linear Formula NH4CoPO4·H2O
Formula Weight 189.96 (171.94 Anhydrous)
MDL Number MFCD02683114
Merck Index 14,513
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2439318

  SDS
alfa aesar™
B2439318
50g Each for $59.38
Description & Specifications

Specifications

Formula Weight 189.96 (171.94 Anhydrous)
CAS 16827-96-6
Health Hazard 1 Danger
Linear Formula NH4CoPO4·H2O
Merck Index 14,513
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H301-H332-H317
Toxic if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Chemical Name or Material Ammonium cobalt(II) phosphate monohydrate
Assay 98%
MDL Number MFCD02683114
TSCA TSCA
Molecular Formula H4CoNO4P·H2O
Quantity 50g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H332-H317
Toxic if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.