missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Citrate Dibasic (Crystalline/Certified ACS), Fisher Chemical™

$315.46 - $554.90

Chemical Identifiers

CAS 3012-65-5
Molecular Formula C6H14N2O7
Molecular Weight (g/mol) 226.185
MDL Number MFCD00013068
InChI Key YXVFQADLFFNVDS-UHFFFAOYSA-N
Synonym ammonium citrate dibasic, ammonium citrate dibasic, puriss, ammonium citrate, dibasic, acs, ammonium hydrogencitrate, citric acid ammonium salt, citric acid diamine, citric acid diammonium salt, citro diamine, di-ammonium hydrogen citrate, diammonium hydrogen citrate
PubChem CID 13710713
ChEBI CHEBI:63076
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A663-500
SDS View Documents
Fisher Chemical
FLA663500
500g
Each for $315.46
Case of 6 Each for $1,892.76
 
A663-212
SDS View Documents
Fisher Chemical
FLA663212
2.5kg
Each for $554.90
Case of 4 Each for $2,219.60
 
Description

Description

Specifications

Chemical Identifiers

3012-65-5
226.185
YXVFQADLFFNVDS-UHFFFAOYSA-N
13710713
azane;2-hydroxypropane-1,2,3-tricarboxylic acid
C6H14N2O7
MFCD00013068
ammonium citrate dibasic, ammonium citrate dibasic, puriss, ammonium citrate, dibasic, acs, ammonium hydrogencitrate, citric acid ammonium salt, citric acid diamine, citric acid diammonium salt, citro diamine, di-ammonium hydrogen citrate, diammonium hydrogen citrate
CHEBI:63076
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.N

Specifications

3012-65-5
C6H14N2O7
MFCD00013068
YXVFQADLFFNVDS-UHFFFAOYSA-N
azane;2-hydroxypropane-1,2,3-tricarboxylic acid
13710713
226.19
98.0 to 103.0%
0.001% max.
5ppm max.
0.01% max.
0.001% max.
Glass Bottle
0.005% max.
4.8 to 5.3
Ammonium Citrate Dibasic
98.0 to 103.0%
(NH4)2O2CCH2C(OH)(CO2H)CH2CO2
ammonium citrate dibasic, ammonium citrate dibasic, puriss, ammonium citrate, dibasic, acs, ammonium hydrogencitrate, citric acid ammonium salt, citric acid diamine, citric acid diammonium salt, citro diamine, di-ammonium hydrogen citrate, diammonium hydrogen citrate
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.N
226.185
CHEBI:63076
Solid
Certified ACS
1.5g/cm³
Pass Test
0.005% max.
P.T. (about 0.05%)
5ppm max.
White
500g
Safety and Handling

Safety and Handling

SDS