Learn More
Ammonium Citrate Dibasic, 98%, ACROS Organics™
Manufacturer: Fisher Scientific Acros 611165000
Specifications
Ammonium citrate dibasic | |
98% | |
MFCD00013068 | |
ammonium citrate dibasic, ammonium citrate dibasic, puriss, ammonium citrate, dibasic, acs, ammonium hydrogencitrate, citric acid ammonium salt, citric acid diamine, citric acid diammonium salt, citro diamine, di-ammonium hydrogen citrate, diammonium hydrogen citrate | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.N | |
226.185 | |
CHEBI:63076 | |
98.0 to 103.0% | |
5ppm max. | |
0.005% max. | |
P.T. (ABOUT 0.05%) | |
0.01% max. | |
500g |
3012-65-5 | |
C6H14N2O7 | |
15, 509 | |
YXVFQADLFFNVDS-UHFFFAOYSA-N | |
azane;2-hydroxypropane-1,2,3-tricarboxylic acid | |
13710713 | |
226.19 | |
0.001% max. | |
Pass Test | |
0.001% max. | |
5ppm max. | |
0.005% max. |
Chemical Identifiers
3012-65-5 | |
226.185 | |
YXVFQADLFFNVDS-UHFFFAOYSA-N | |
13710713 | |
azane;2-hydroxypropane-1,2,3-tricarboxylic acid |
C6H14N2O7 | |
MFCD00013068 | |
ammonium citrate dibasic, ammonium citrate dibasic, puriss, ammonium citrate, dibasic, acs, ammonium hydrogencitrate, citric acid ammonium salt, citric acid diamine, citric acid diammonium salt, citro diamine, di-ammonium hydrogen citrate, diammonium hydrogen citrate | |
CHEBI:63076 | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.N |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 221-146-3
TSCA : TSCA