Learn More
Ammonium Citrate Dibasic, 98%, Thermo Scientific Chemicals
Supplier: Fisher Scientific Acros 611160025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium citrate dibasic | |
2.5kg | |
C6H14N2O7 | |
15, 509 | |
YXVFQADLFFNVDS-UHFFFAOYSA-N | |
diammonium 3-carboxy-3-hydroxypentanedioate | |
13710713 | |
226.19 | |
5ppm max. | |
0.005% max. | |
P.T. (ABOUT 0.05%) | |
5ppm max. | |
0.005% max. |
3012-65-5 | |
98% | |
MFCD00013068 | |
ammonium citrate dibasic, ammonium citrate dibasic, puriss, ammonium citrate, dibasic, acs, ammonium hydrogencitrate, citric acid ammonium salt, citric acid diamine, citric acid diammonium salt, citro diamine, di-ammonium hydrogen citrate, diammonium hydrogen citrate | |
[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O | |
226.19 | |
CHEBI:63076 | |
98.0 to 103.0% | |
Pass Test | |
0.001% max. | |
Poly Pail | |
0.01% max. | |
0.001% max. |
Chemical Identifiers
3012-65-5 | |
226.19 | |
YXVFQADLFFNVDS-UHFFFAOYSA-N | |
13710713 | |
diammonium 3-carboxy-3-hydroxypentanedioate |
C6H14N2O7 | |
MFCD00013068 | |
ammonium citrate dibasic, ammonium citrate dibasic, puriss, ammonium citrate, dibasic, acs, ammonium hydrogencitrate, citric acid ammonium salt, citric acid diamine, citric acid diammonium salt, citro diamine, di-ammonium hydrogen citrate, diammonium hydrogen citrate | |
CHEBI:63076 | |
[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 221-146-3
TSCA : TSCA
RUO – Research Use Only