Learn More
Ammonium Citrate Dibasic, 98%, ACROS Organics™
$130.49 - $341.65
Chemical Identifiers
CAS | 3012-65-5 |
---|---|
Molecular Formula | C6H14N2O7 |
Molecular Weight (g/mol) | 226.185 |
MDL Number | MFCD00013068 |
InChI Key | YXVFQADLFFNVDS-UHFFFAOYSA-N |
Synonym | ammonium citrate dibasic, ammonium citrate dibasic, puriss, ammonium citrate, dibasic, acs, ammonium hydrogencitrate, citric acid ammonium salt, citric acid diamine, citric acid diammonium salt, citro diamine, di-ammonium hydrogen citrate, diammonium hydrogen citrate |
PubChem CID | 13710713 |
ChEBI | CHEBI:63076 |
IUPAC Name | azane;2-hydroxypropane-1,2,3-tricarboxylic acid |
SMILES | C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC611165000
|
Fisher Scientific Acros
611165000 |
500g |
Each for $130.49
|
|
AC611160025
|
Fisher Scientific Acros
611160025 |
2.5kg |
Each for $341.65
|
|
Chemical Identifiers
3012-65-5 | |
226.185 | |
YXVFQADLFFNVDS-UHFFFAOYSA-N | |
13710713 | |
azane;2-hydroxypropane-1,2,3-tricarboxylic acid |
C6H14N2O7 | |
MFCD00013068 | |
ammonium citrate dibasic, ammonium citrate dibasic, puriss, ammonium citrate, dibasic, acs, ammonium hydrogencitrate, citric acid ammonium salt, citric acid diamine, citric acid diammonium salt, citro diamine, di-ammonium hydrogen citrate, diammonium hydrogen citrate | |
CHEBI:63076 | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.N |
Specifications
3012-65-5 | |
C6H14N2O7 | |
15, 509 | |
YXVFQADLFFNVDS-UHFFFAOYSA-N | |
azane;2-hydroxypropane-1,2,3-tricarboxylic acid | |
13710713 | |
226.19 | |
0.001% max. | |
Pass Test | |
0.001% max. | |
5ppm max. | |
0.005% max. | |
Ammonium citrate dibasic |
98% | |
MFCD00013068 | |
ammonium citrate dibasic, ammonium citrate dibasic, puriss, ammonium citrate, dibasic, acs, ammonium hydrogencitrate, citric acid ammonium salt, citric acid diamine, citric acid diammonium salt, citro diamine, di-ammonium hydrogen citrate, diammonium hydrogen citrate | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.N | |
226.185 | |
CHEBI:63076 | |
98.0 to 103.0% | |
5ppm max. | |
0.005% max. | |
P.T. (ABOUT 0.05%) | |
0.01% max. | |
500g |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 221-146-3
TSCA : TSCA