Please login to your online account to display your discounted pricing

Ammonium chromate, Reagent Grade, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Ammonium chromate
CAS 7788-98-9
Purity Grade Reagent Grade
Physical Form Crystalline
Molecular Formula (NH4)2CrO4
Formula Weight 152.07
MDL Number MFCD00015958
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA8768022

 
alfa aesar
8768022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA8768036

 
alfa aesar
8768036
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Reagent Grade
Formula Weight 152.07
Quantity 100g
Color Yellow-orange
DOT Information DOT Class: 5.1, Packaging Group: III
EINECS Number 232-138-4
Physical Form Crystalline
Density 1.91g/cm3
Chemical Name or Material Ammonium chromate
Odor Odorless
Molecular Formula (NH4)2CrO4
Appearance Yellow-orange crystalline
MDL Number MFCD00015958
CAS 7788-98-9
Health Hazard 1 H350-H400-H410-H317
Safety and Handling

H350-H400-H410-H317