missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Ammonium Chloride, Reagent Grade, ≥99.5%, Honeywell™

Reagent Grade,≥99.5%

$227.67 - $227.67

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.489
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name azanium;chloride
SMILES [NH4+].[Cl-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60047821 View Documents Honeywell™
A45141KG
1kg Poly Bottle Each for $227.67
 
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

12125-02-9
53.489
NLXLAEXVIDQMFP-UHFFFAOYSA-N
25517
azanium;chloride
ClH4N
MFCD00011420
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
CHEBI:31206
[NH4+].[Cl-]

Specifications

12125-02-9
NH4Cl
4371014
NLXLAEXVIDQMFP-UHFFFAOYSA-N
azanium;chloride
25517
53.49g/mol
1.536g/cm3 at 20°C
Poly Bottle
1.9 (vs air)
Colorless
4.6 to 6 (at 20°C)
Ammonium chloride
ClH4N
MFCD00011420
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
[NH4+].[Cl-]
53.489
CHEBI:31206
≥99.5%
Odorless
1mmHg (160.4°C)
Solid
338°C
1kg
Safety and Handling

Safety and Handling

P305 + P351 + P338

RTECSNumber : 235-186-4