Please login to your online account to display your discounted pricing

Ammonium Chloride, Molecular Biology Grade, Calbiochem™, EMD Millipore

$48.75 - $73.50

Chemical Identifiers

Assay Percent Range ≥99%
Formula Weight 53.5
CAS 12125-02-9
Chemical Name or Material Ammonium Chloride
Density 1.53g/cm3 at 25°C
Molecular Formula NH4Cl
Purity Grade Notes by argentometry
Solubility Information Easily soluble in water
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.
Synonym Amchlor; Ammoneric; Ammonium Muriate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

1683201KG

 
emd millipore corporation
1683201KG
Each for $73.50

168320250GM

 
emd millipore corporation
168320250GM
Each for $48.75
Description & Specifications

Specifications

Formula Weight 53.5
Quantity 1kg
Chemical Name or Material Ammonium Chloride
Purity Grade Notes by argentometry
Assay Percent Range ≥99%
Molecular Formula NH4Cl
CAS 12125-02-9
RNase None detected
Synonym Amchlor; Ammoneric; Ammonium Muriate
Solubility Information Easily soluble in water
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.
Density 1.53g/cm3 at 25°C
DNase None detected
RTECS Number BP4550000
Safety and Handling

Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.