Please login to your online account to display your discounted pricing

Ammonium Chloride, For Nitrogen (Nitrate), Certified, 85g/L, LabChem™

$50.14 - $78.94

Chemical Identifiers

Chemical Name or Material Ammonium Chloride
CAS 7732-18-5
CAS Max % 91.45
CAS 12125-02-9
CAS Max % 8.5
CAS 9002-92-0
CAS Max % 0.05
Name Note For Nitrogen (Nitrate)
Purity Grade Certified Grade
Linear Formula NH4Cl
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC109901

 
lab chem
LC109901
Poly Bottle 500mL Each for $50.14

LC109902

 
lab chem
LC109902
Poly Bottle 1L Each for $78.94
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 53.49
Solubility Information Soluble in water
Name Note For Nitrogen (Nitrate)
CAS 9002-92-0
Physical Form Liquid
CAS Max % 91.45
Quantity 500mL
Identification Passes Test
pH 4.0 to 5.0
CAS Max % 8.5
Concentration 85g/L
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
CAS Max % 0.05
CAS 7732-18-5
Chemical Name or Material Ammonium Chloride
Linear Formula NH4Cl
Color Clear, colorless
Recommended Storage Room Temperature
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 12125-02-9
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature