Please login to your online account to display your discounted pricing

Ammonium Chloride, For BOD, Certified, 1.15g/L, LabChem™

$39.57 - $39.57

Chemical Identifiers

Chemical Name or Material Ammonium Chloride
CAS 7732-18-5
CAS Max % 99.8847
CAS 12125-02-9
CAS Max % 0.115
CAS 1310-73-2
CAS Max % 0.0003
Name Note For BOD
Purity Grade Certified Grade
Linear Formula NH4Cl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC109801

 
lab chem
LC109801
500mL Each for $39.57
Description & Specifications

Specifications

Formula Weight 53.49
pH 7.20 ± 0.05
Purity Grade Certified Grade
Solubility Soluble in water
Physical Form Liquid
Quantity 500mL
CAS 7732-18-5
Storage Information Room Temperature
Chemical Name or Material Ammonium Chloride
Concentration 1.15g/L
CAS 12125-02-9
Packaging Poly Bottle
Name Note For BOD
CAS 1310-73-2
Decomposition Information Gaseous Ammonia
Assay Percent Notes in-process pH
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
CAS Max % 0.115
CAS Max % 0.0003
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 99.8847
Color Clear, colorless
Linear Formula NH4Cl
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.