missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium Chloride-EDTA Solution, Ricca Chemical
For Nitrate by the Cadmium Reduction Method
$259.38 - $259.38
Chemical Identifiers
CAS | 12125-02-9 |
---|---|
Molecular Formula | ClH4N |
Molecular Weight (g/mol) | 53.49 |
MDL Number | MFCD00011420 |
InChI Key | NLXLAEXVIDQMFP-UHFFFAOYSA-N |
Synonym | ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid |
PubChem CID | 25517 |
ChEBI | CHEBI:31206 |
IUPAC Name | amine hydrochloride |
SMILES | N.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
626-32
|
Ricca Chemical Company
62632 |
1 L | Natural Poly Bottle |
Each for $259.38
|
|
||||
Chemical Identifiers
12125-02-9 | |
53.49 | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
25517 | |
amine hydrochloride |
ClH4N | |
MFCD00011420 | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
CHEBI:31206 | |
N.Cl |
Specifications
Colorless | |
ClH4N | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
N.Cl | |
53.49 | |
CHEBI:31206 | |
1 L | |
Ammonium Chloride-EDTA Solution |
12125-02-9,1336-21-6,7732-18-5 | |
MFCD00011420 | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
amine hydrochloride | |
25517 | |
Natural Poly Bottle | |
Liquid |