Please login to your online account to display your discounted pricing

Ammonium Chloride, Certified, 10.0% (w/v) ± 0.5%, LabChem™

$81.03 - $81.03

Chemical Identifiers

Chemical Name or Material Ammonium Chloride
CAS 7732-18-5
CAS Max % 90
CAS 12125-02-9
CAS Max % 10
Purity Grade Certified Grade
Linear Formula NH4Cl
Formula Weight 53.49
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC110102

 
lab chem
LC110102
1L Each for $81.03
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 53.49
Assay 10.0% ± 0.5% (w/v)
Viscosity 0.94cSt
Identification Passes Test
Health Hazard 2 GHS H Statement
Causes serious eye irritation.
Decomposition Information Gaseous Ammonia
CAS Max % 90
Density 1.03g/mL
Linear Formula NH4Cl
CAS 7732-18-5
CAS 12125-02-9
CAS Max % 10
Chemical Name or Material Ammonium Chloride
Quantity 1L
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Physical Form Liquid
Solubility Soluble in water
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Color Clear, colorless
Specific Gravity 1.03g/mL
Storage Information Room Temperature
Safety and Handling

Warning

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes serious eye irritation.