Please login to your online account to display your discounted pricing

Ammonium Chloride, Certified, 0.1000M ± 0.0005M (0.1N), LabChem™

$63.03 - $63.03

Chemical Identifiers

Chemical Name or Material Ammonium Chloride
CAS 7732-18-5
CAS Max % 99.47
CAS 12125-02-9
CAS Max % 0.53
Purity Grade Certified Grade
Linear Formula NH4Cl
Formula Weight 53.49
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC109751

 
lab chem
LC109751
500mL Each for $63.03
Description & Specifications

Specifications

Formula Weight 53.49
Purity Grade Certified Grade
Quantity 500mL
CAS 7732-18-5
Color Clear, colorless
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Concentration 0.1000M ± 0.0005M (0.1N)
Packaging Poly Bottle
Density 1g/mL
Physical Form Liquid
Linear Formula NH4Cl
Viscosity 1cSt
CAS Max % 0.53
Chemical Name or Material Ammonium Chloride
CAS Max % 99.47
Identification Passes Test
Decomposition Information Gaseous Ammonia
Storage Information Room Temperature
Specific Gravity 1g/mL
CAS 12125-02-9
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.