Please login to your online account to display your discounted pricing

Ammonium Chloride, ACS Grade, ≥ 99.5%, LabChem™

$56.49 - $56.49

Chemical Identifiers

Chemical Name or Material Ammonium Chloride
CAS 12125-02-9
CAS Max % 100
Purity Grade ACS Grade
Linear Formula NH4Cl
Formula Weight 53.49
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC109721

 
lab chem
LC109721
500g Each for $56.49
Description & Specifications

Specifications

Assay ≥99.5%
Formula Weight 53.49
Physical Form Free-Flowing Crystals
Linear Formula NH4Cl
Storage Information Room Temperature
Purity Grade ACS Grade
Health Hazard 3 GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.
CAS 12125-02-9
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Identification Passes Test
Solubility Soluble in water
Color White
Density 1530kg/m3
EINECS Number 235-186-4
Packaging Poly Bottle
Quantity 500g
CAS Max % 100
Chemical Name or Material Ammonium Chloride
Specific Gravity 1530kg/m3
Health Hazard 1 Warning
Decomposition Information Gaseous Ammonia
Safety and Handling

Warning

GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.