missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium chloride, 99+%, pure, Thermo Scientific Chemicals

Ammonium chloride, 99+%, H4ClN, CAS Number-12125-02-9, amchlor, ammoneric, ammoniumchlorid, ammonium chloride, ammonium chloride nh4 cl, ammoniumklorid, ammonium muriate, darammon, sal ammoniac, salmiac, 1kg, 99% min. (Argentometry), CHEBI:31206, White, 1.5200g/mL, 235-186-4, 15,13, 53.49, Pure

Supplier:  Thermo Scientific Chemicals 123340010

 View more versions of this product

Catalog No. AC123340010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium chloride
12125-02-9
Crystalline Powder and/or Chunks
99% min. (Argentometry)
NH3·HCl
15,13
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
NLXLAEXVIDQMFP-UHFFFAOYSA-N
azanium;chloride
25517
53.49
Pure
1.5200g/mL
340.0°C
White
1 kg
ClH4N
MFCD00011420
15, 506
Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol,practically insoluble in acetone,ether and,ethyl acetate
N.Cl
53.49
CHEBI:31206
≥99%
Plastic Bottle

Chemical Identifiers

12125-02-9
53.49
NLXLAEXVIDQMFP-UHFFFAOYSA-N
25517
N.Cl
ClH4N
MFCD00011420
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
CHEBI:31206
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not eat,d

GHS Signal Word: Warning

EINECSNumber : 235-186-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only