missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium chloride, 99.5% min, Alfa Aesar

Manufacturer:  Thermo Fisher Scientific 12361A3

 View more versions of this product

Catalog No. AA12361A3



Specifications

Specifications

Ammonium chloride
99.5% min.
MFCD00011420
amchlor
NLXLAEXVIDQMFP-UHFFFAOYSA-N
azanium;chloride
25517
53.49
1.53
1.642
340° (sublimation)
12125-02-9
ClH4N
Granular
Soluble in liquid ammonia,acetone,hydrazine,alcohol. Insoluble in diethyl ether,ethyl acetate.
[NH4+].[Cl-]
53.489
CHEBI:31206
Granular
Odorless
Hygroscopic
2kg

Chemical Identifiers

12125-02-9
53.489
NLXLAEXVIDQMFP-UHFFFAOYSA-N
25517
azanium;chloride
ClH4N
MFCD00011420
amchlor
CHEBI:31206
[NH4+].[Cl-]
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P314-P330-P501c

H302-H319-H335-H373

EINECSNumber : 235-186-4

RTECSNumber : BP4550000

TSCA : Yes

storageNote1 : Hygroscopic

Recommended Storage : Ambient temperatures

Documents