Learn More
Ammonium chloride, >=99.0%, extra pure, with anti-caking agent, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 396400050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ammonium chloride | |
5kg | |
H4ClN | |
MFCD00011420 | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
azanium;chloride | |
25517 | |
53.49 | |
Extra Pure | |
1.52 |
12125-02-9,7732-18-5 | |
99.0 to 101% (Argentometry) | |
NH4Cl | |
15, 506 | |
Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol,practically insoluble in acetone,ether and ethyl,acetate | |
[NH4+].[Cl-] | |
53.49 | |
CHEBI:31206 | |
≥99% | |
Plastic drum | |
1.5200g/mL |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wash face,ha
GHS Signal Word: Warning
EINECSNumber : 235-186-4
RUO â Research Use Only