missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Chloride, ∽98%, MP Biomedicals

Manufacturer:  Mp Biomedicals 19480691

 View more versions of this product

Catalog No. ICN19480691


 

Specifications Description & Specifications

Specifications

Ammonium Chloride
∽98%
53.5
12125-02-9
NH4Cl
1kg

Chemical Identifiers

∽98%
NH4Cl
53.5
Ammonium Chloride

Safety and Handling

Recommended Storage : Room Temperature