Please login to your online account to display your discounted pricing

Ammonium Chloride, ∽98%, MP Biomedicals


Quantity 500g

Manufacturer: mp biomedicals  19480690

 View more versions of this product

Catalog No. ICN19480690

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 53.5
Quantity 500g
CAS 12125-02-9
Molecular Formula NH4Cl
Assay Percent Range ∽98%
Recommended Storage Room Temperature
Chemical Name or Material Ammonium Chloride

Specifications

Chemical Name or Material Ammonium Chloride
Assay Percent Range ∽98%
Molecular Formula NH4Cl
Formula Weight 53.5
Accessories

Room Temperature