Please login to your online account to display your discounted pricing

Ammonium Chloride, ∽98%, MP Biomedicals

$53.21 - $101.78

Chemical Identifiers

Chemical Name or Material Ammonium Chloride
Assay Percent Range ∽98%
Molecular Formula NH4Cl
Formula Weight 53.5
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19480680

 
mp biomedicals
19480680
100g Each for $53.21

ICN19480690

 
mp biomedicals
19480690
500g Each for $75.06

ICN19480691

 
mp biomedicals
19480691
1kg Each for $101.78
Description & Specifications

Specifications

Formula Weight 53.5
Quantity 100g
Chemical Name or Material Ammonium Chloride
Assay Percent Range ∽98%
Molecular Formula NH4Cl
CAS 12125-02-9
Recommended Storage Room Temperature
Safety and Handling

Room Temperature