Learn More
Ammonium chloride, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1500030
Description
Ammonium Chloride is commonly used as a buffer solution. It is used in the manufacture of fireworks, safety explosives and smokes. It is used to control pH levels in chemicals, to produce sodium carbonate, as a fertilizer, in electronics and in metallurgy. Also used as a flux for cleaning, as a flavoring agent, as an expectorant in cough medicine and as a systemic acidifying agent. It is widely used as an electrolyte in zinc-carbon batteries. It has application in leather industries for dyeing, tanning, textile printing and to luster cotton.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAmmonium Chloride is commonly used as a buffer solution. It is used in the manufacture of fireworks, safety explosives and smokes. It is used to control pH levels in chemicals, to produce sodium carbonate, as a fertilizer, in electronics and in metallurgy. Also used as a flux for cleaning, as a flavoring agent, as an expectorant in cough medicine and as a systemic acidifying agent. It is widely used as an electrolyte in zinc-carbon batteries. It has application in leather industries for dyeing, tanning, textile printing and to luster cotton.
Solubility
Soluble in water, methanol, liquid ammonia and acetone. Insoluble in diethyl ether and ethyl acetate.
Notes
Hygroscopic. Incompatible with silver nitrate and strong oxidizers.
Specifications
Ammonium chloride | |
12125-02-9 | |
ClH4N | |
MFCD00011420 | |
14,509 | |
Soluble in water,methanol,liquid ammonia and acetone. Insoluble in diethyl ether and ethyl acetate. | |
N.Cl | |
53.49 | |
CHEBI:31206 | |
≥98% | |
1.642 | |
1.53 |
340°C (sublimation) | |
250 g | |
NH4Cl | |
4371014 | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
azanium;chloride | |
25517 | |
53.49 | |
Odorless | |
Hygroscopic |
Chemical Identifiers
12125-02-9 | |
53.49 | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
25517 | |
N.Cl |
ClH4N | |
MFCD00011420 | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
CHEBI:31206 |
Safety and Handling
GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P314-P330-P501c
H302-H319-H335-H373
EINECSNumber : 235-186-4
RTECSNumber : BP4550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only