missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Chloride, 8.5% (w/v) Aqueous Solution, with Brij 35, Ricca Chemical

$76.55 - $76.55

Chemical Identifiers

CAS 7732-18-5,12125-02-9,9002-92-0
CAS Min % 90.0,8.0,0.05
CAS Max % 94.0,9.0,0.05
Physical Form Liquid
Chemical Name or Material Ammonium Chloride, With Brij 35
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
6252-16 Ricca Chemical Company
RI625.216
Poly natural 500mL Each for $76.55
6252-32 Ricca Chemical Company
RI625.232
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,12125-02-9,9002-92-0
94.0,9.0,0.05
Ammonium Chloride, With Brij 35
90.0,8.0,0.05
Liquid

Specifications

7732-18-5,12125-02-9,9002-92-0
94.0,9.0,0.05
Poly natural
Aqueous Solution
500mL
90.0,8.0,0.05
Liquid
8.5% (w/v)
Clear colorless
Ammonium Chloride, With Brij 35