Please login to your online account to display your discounted pricing

Ammonium Chloride, 5% (w/v), LabChem™

$80.46 - $80.46

Chemical Identifiers

Chemical Name or Material Ammonium Chloride
CAS 7732-18-5
CAS Max % 95
CAS 12125-02-9
CAS Max % 5
Linear Formula NH4Cl
Formula Weight 53.49
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC110002

 
lab chem
LC110002
1L Each for $80.46
Description & Specifications

Specifications

Formula Weight 53.49
Chemical Name or Material Ammonium Chloride
Identification Passes Test
Packaging Poly Bottle
Physical Form Liquid
Assay 5% (w/v)
Solubility Soluble in water
Specific Gravity 1.01g/mL
Storage Information Room Temperature
Linear Formula NH4Cl
CAS 7732-18-5
Viscosity 0.97cSt
Decomposition Information Gaseous Ammonia
Color Clear, colorless
Quantity 1L
CAS Max % 95
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 12125-02-9
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 5
Density 1.01g/mL
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.