missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Chloride, 5% (w/v) Aqueous Solution (50g/L), Ricca Chemical

$58.67 - $58.67

Chemical Identifiers

CAS 7732-18-5,12125-02-9
CAS Min % 93.0,4.0
CAS Max % 97.0,6.0
Physical Form Liquid
Chemical Name or Material Ammonium Chloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
625-16 Ricca Chemical Company
RI62516
Poly natural 500mL Each for $58.67
62532 Ricca Chemical Company
RI62532
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R06250004A Ricca Chemical Company
RIR06250004A
Poly natural 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,12125-02-9
97.0,6.0
Ammonium Chloride
93.0,4.0
Liquid

Specifications

7732-18-5,12125-02-9
97.0,6.0
Poly natural
Aqueous Solution
500mL
93.0,4.0
Liquid
5% (w/v),(50g/L)
Clear colorless
Ammonium Chloride