missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium Chloride, 5% (w/v) (50 g/L), Ricca Chemical
Supplier: Ricca Chemical Company 62516
Specifications
Ammonium Chloride | |
12125-02-9,7732-18-5 | |
97.0,6.0 | |
MFCD00011420 | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
amine hydrochloride | |
25517 | |
Natural Poly Bottle | |
500 mL | |
5% (w/v),(50g/L) |
Colorless | |
93.0,4.0 | |
ClH4N | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
N.Cl | |
53.49 | |
CHEBI:31206 | |
Aqueous Solution | |
Liquid |
Chemical Identifiers
12125-02-9 | |
53.49 | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
25517 | |
amine hydrochloride |
ClH4N | |
MFCD00011420 | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
CHEBI:31206 | |
N.Cl |