Please login to your online account to display your discounted pricing

Ammonium Chloride, 10% (w/v) Aqueous Solution, Ricca Chemical

$131.55 - $131.55

Chemical Identifiers

Chemical Name or Material Ammonium Chloride
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 93.0
CAS 12125-02-9
CAS Min % 9.0
CAS Max % 11.0
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R06254004A

 
ricca chemical company
RIR06254004A
Poly natural 4L Each for $131.55

6254-16

 
ricca chemical company
RI625.416
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

6254-32

 
ricca chemical company
RI625.432
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R062540020F

 
ricca chemical company
RIR062540020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
CAS 7732-18-5
CAS Min % 89.0
Solvent or Matrix Aqueous Solution
Concentration or Composition Notes 10% (w/v)
Packaging Poly natural
Chemical Name or Material Ammonium Chloride
CAS 12125-02-9
CAS Max % 93.0
CAS Min % 9.0
CAS Max % 11.0