missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Chloride, 10% (w/v) Aqueous Solution, Ricca Chemical

Manufacturer:  Ricca Chemical Company RIR062540020F

 View more versions of this product

Catalog No. R062540020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ammonium Chloride
89.0,9.0
ClH4N
NLXLAEXVIDQMFP-UHFFFAOYSA-N
azanium;chloride
25517
Liquid
10% (w/v)
Colorless
12125-02-9,7732-18-5
93.0,11.0
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
[NH4+].[Cl-]
53.489
CHEBI:31206
Cubitainer
Aqueous Solution
20L

Chemical Identifiers

12125-02-9
53.489
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
CHEBI:31206
[NH4+].[Cl-]
ClH4N
NLXLAEXVIDQMFP-UHFFFAOYSA-N
25517
azanium;chloride
Documents