Please login to your online account to display your discounted pricing

Ammonium Chloride, 1.15g/L, pH 7.20, for BOD, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Ammonium Chloride
Name Note pH 7.20, for BOD
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 12125-02-9
CAS Min % 0.11
CAS Max % 0.12
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

623-16

 
ricca chemical company
RI62316
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

623-4

 
ricca chemical company
RI6234
Poly natural 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 500mL
Chemical Name or Material Ammonium Chloride
Name Note pH 7.20, for BOD
Concentration or Composition Notes 1.15g/L
CAS Min % 98.0
CAS Min % 0.11
Packaging Poly natural
CAS 7732-18-5
CAS Max % 100.0
CAS 12125-02-9
CAS Max % 0.12