missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium chloride, 0.5M aq. soln.

$50.74 - $75.88

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.489
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name azanium;chloride
SMILES [NH4+].[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62046AD View Documents Alfa Aesar™
J62046AD
50mL Each for $50.74
 
AAJ62046AE View Documents Alfa Aesar™
J62046AE
100mL Each for $75.88
 
Description

Description

Applications
Ammonium chloride is used for the lysis of human red blood cells. It is used in protein isolation techniques and in the study of basic calcium phosphate crystals in fibroblasts. It inhibits intracellular dissolution of basic calcium phosphate (BCP) crystals by alkalinizing phagolysosomes. It is used as a flavoring agent in some types of liquorices. It is also used as electrolyte in zinc-carbon batteries.

Solubility
Fully miscible in water. Soluble in ethanol, methanol, and glycerol. Insoluble in acetone, and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

12125-02-9
53.489
NLXLAEXVIDQMFP-UHFFFAOYSA-N
25517
azanium;chloride
ClH4N
MFCD00011420
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
CHEBI:31206
[NH4+].[Cl-]

Specifications

12125-02-9
NH4Cl
4371014
NLXLAEXVIDQMFP-UHFFFAOYSA-N
azanium;chloride
25517
53.49
0.5M aq. soln.
50mL
ClH4N
MFCD00011420
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
[NH4+].[Cl-]
53.489
CHEBI:31206
Liquid
Colorless
Ammonium chloride
Safety and Handling

Safety and Handling

RTECSNumber : BP4550000

TSCA : TSCA