missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium chloride, 0.5M aq. soln., Thermo Scientific™

Supplier:  Thermo Scientific Chemicals J62046AD

 View more versions of this product

Catalog No. AAJ62046AD



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ammonium chloride is used for the lysis of human red blood cells. It is used in protein isolation techniques and in the study of basic calcium phosphate crystals in fibroblasts. It inhibits intracellular dissolution of basic calcium phosphate (BCP) crystals by alkalinizing phagolysosomes. It is used as a flavoring agent in some types of liquorices. It is also used as electrolyte in zinc-carbon batteries.

Solubility
Fully miscible in water. Soluble in ethanol, methanol, and glycerol. Insoluble in acetone, and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

Ammonium chloride
12125-02-9
50 mL
NH4Cl
4371014
Fully miscible in water. Soluble in ethanol,methanol,and glycerol. Insoluble in acetone,and ether.
N.Cl
53.49
CHEBI:31206
0.5M aq. soln.
Colorless
Liquid
ClH4N
MFCD00011420
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
NLXLAEXVIDQMFP-UHFFFAOYSA-N
azanium;chloride
25517
53.49

Chemical Identifiers

12125-02-9
53.49
NLXLAEXVIDQMFP-UHFFFAOYSA-N
25517
azanium;chloride
ClH4N
MFCD00011420
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
CHEBI:31206
N.Cl
Safety and Handling

Safety and Handling

EINECSNumber : 235-186-4

RTECSNumber : BP4550000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions