missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Chloride, 0.100 Molar, Ricca Chemical

$75.58 - $75.58

Chemical Identifiers

CAS 12125-02-9 (Ammonium Chloride),7732-18-5 (Water, Deionized)
Physical Form Liquid
Chemical Name or Material Ammonium Chloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
627-16 Ricca Chemical Company
RI62716
Poly natural 500mL Each for $75.58
R06270001A Ricca Chemical Company
RIR06270001A
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

12125-02-9 (Ammonium Chloride),7732-18-5 (Water, Deionized)
Liquid
Ammonium Chloride

Specifications

Liquid
500mL
7732-18-5 (Water, Deionized),12125-02-9 (Ammonium Chloride)
Ammonium Chloride
Clear colorless
0.100Molar
Poly natural