Please login to your online account to display your discounted pricing

Ammonium Cerium(IV)sulfate Dihydrate, Reagent ACS 94%, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC325790025

 View more versions of this product

Catalog No. AC325790025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Ammonium cerium(IV) sulfate dihydrate
CAS 10378-47-9
Purity Grade ACS Reagent Grade
Physical Form Crystalline Powder
Assay ≥94%
Assay Percent Range 95%
Molecular Formula H16CeN4O16S4·2H2O
Linear Formula (NH4)4Ce(SO4)4·2H2O
Formula Weight 632.54
Fieser 08,80; 01,1168; 02,414; 11,519; 12,486; 13,298; 17,333
Synonym Ceric ammonium sulfate
Insoluble Matter 0.05% (in dilute H2SO4) max.
Iron (Fe) 100ppm max.
Packaging Plastic bottle
Phosphate 300ppm max.
Color Orange
Quantity 2.5kg

Specifications

Chemical Name or Material Ammonium cerium(IV) sulfate dihydrate
CAS 10378-47-9
Purity Grade ACS Reagent Grade
Assay ≥94%
Assay Percent Range 95%
Molecular Formula H16CeN4O16S4·2H2O
Linear Formula (NH4)4Ce(SO4)4·2H2O
Formula Weight 632.54
Synonym Ceric ammonium sulfate