missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium cerium(IV) sulfate dihydrate, 95%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 325795000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium cerium(IV) sulfate dihydrate | |
Orange | |
Crystalline Powder | |
95% | |
(NH4)4Ce(SO4)4·2H2O | |
Ceric ammonium sulfate | |
632.54 | |
95% | |
Plastic bottle |
130.0°C | |
10378-47-9 | |
500 g | |
H16CeN4O16S4·2H2O | |
08,8 | |
Solubility in water: hydrolysis. Other solubilities: insoluble in ethanol | |
632.54 | |
ACS Reagent |
Chemical Identifiers
10378-47-9 | |
H16CeN4O16S4·2H2O | |
632.54 | |
Ceric ammonium sulfate |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only