missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium cerium(IV) sulfate dihydrate, 99+%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 206315000

 View more versions of this product

Catalog No. AC206315000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium cerium(IV) sulfate dihydrate
Extra Pure
10378-47-9
500 g
H16CeN4O16S4·2H2O
MFCD00149417
Ceric ammonium sulfate
632.54
≥99%
Plastic bottle
130.0°C
Orange
Crystalline Powder
99+%
(NH4)4Ce(SO4)4·2H2O
08,80
Solubility in water: hydrolysis. Other solubilities: insoluble in ethanol
632.54
Extra Pure

Chemical Identifiers

10378-47-9
632.54
Ceric ammonium sulfate
H16CeN4O16S4·2H2O
MFCD00149417
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only